Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-nitropyridine, 96%


Quantity 1g

Manufacturer: alfa aesar™  L1972503

 View more versions of this product

Catalog No. AAL1972503

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 139.11
Chemical Name or Material 4-Amino-3-nitropyridine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00160304
Beilstein 383710
Synonym 3-Nitro-4-pyridinamine
Molecular Formula C5H5N3O2
Assay 96%
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Melting Point 201° to 205°C
CAS 1681-37-4

Specifications

Chemical Name or Material 4-Amino-3-nitropyridine
CAS 1681-37-4
Molecular Formula C5H5N3O2
Formula Weight 139.11
MDL Number MFCD00160304
Beilstein 383710
Synonym 3-Nitro-4-pyridinamine

Applications
4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Accessories

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents