missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-nitrophenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444570010

 View more versions of this product

Catalog No. AC444570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Amino-3-nitrophenylboronic acid pinacol ester
174°C to 177°C
833486-94-5
C12H17BN2O4
QOYJKGGBFKVKDP-UHFFFAOYSA-N
16217914
97%
Glass Bottle
1g
96% min. (GC)
2-4-amino-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 4-amino-3-nitrophenylboronic acid pinacol ester, 4-amino-3-nitrophenylboronic acid, pinacol ester, acmc-209ps0, benzenamine, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, ksc912g7t, pubchem22339, qoyjkggbfkvkdp-uhfffaoysa-n
264.09
264.09

Chemical Identifiers

833486-94-5
264.09
2-4-amino-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 4-amino-3-nitrophenylboronic acid pinacol ester, 4-amino-3-nitrophenylboronic acid, pinacol ester, acmc-209ps0, benzenamine, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, ksc912g7t, pubchem22339, qoyjkggbfkvkdp-uhfffaoysa-n
C12H17BN2O4
QOYJKGGBFKVKDP-UHFFFAOYSA-N
16217914
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents