missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-nitrophenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 833486-94-5
Molecular Formula C12H17BN2O4
Molecular Weight (g/mol) 264.09
InChI Key QOYJKGGBFKVKDP-UHFFFAOYSA-N
Synonym 4-amino-3-nitrophenylboronic acid pinacol ester, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-nitrophenylboronic acid, pinacol ester, 2-4-amino-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pubchem22339, acmc-209ps0, ksc912g7t, qoyjkggbfkvkdp-uhfffaoysa-n
PubChem CID 16217914
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444570050 SDS View Documents Acros Organics
AC444570050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444570010 SDS View Documents Acros Organics
AC444570010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

833486-94-5
264.09
4-amino-3-nitrophenylboronic acid pinacol ester, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-nitrophenylboronic acid, pinacol ester, 2-4-amino-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pubchem22339, acmc-209ps0, ksc912g7t, qoyjkggbfkvkdp-uhfffaoysa-n
C12H17BN2O4
QOYJKGGBFKVKDP-UHFFFAOYSA-N
16217914

Specifications

5g
97%
4-amino-3-nitrophenylboronic acid pinacol ester, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-nitrophenylboronic acid, pinacol ester, 2-4-amino-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pubchem22339, acmc-209ps0, ksc912g7t, qoyjkggbfkvkdp-uhfffaoysa-n
264.09
264.09
833486-94-5
C12H17BN2O4
QOYJKGGBFKVKDP-UHFFFAOYSA-N
16217914
4-Amino-3-nitrophenylboronic acid pinacol ester
Safety and Handling

Safety and Handling

Warning