Please login to your online account to display your discounted pricing

4-Amino-3-nitrophenol, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  33522-0250

 View more versions of this product

Catalog No. AC335220250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Melting Point >150°C
Formula Weight 154.13
Quantity 25g
Color Bordeaux to deep red
Beilstein 13, III, 1203
CAS Max % 100.0
Chemical Name or Material 4-Amino-3-nitrophenol
Molecular Formula C6H6N2O3
MDL Number MFCD00066310
CAS 610-81-1
Synonym 2-Amino-5-hydroxynitrobenzene
Assay Percent Range 98%
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause respiratory irritation.
Harmful if swallowed.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Amino-3-nitrophenol
CAS 610-81-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6N2O3
Formula Weight 154.13
MDL Number MFCD00066310
Synonym 2-Amino-5-hydroxynitrobenzene
Accessories

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents