missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-nitrobenzotrifluoride 98%, ACROS Organics™

$69.98 - $69.98

Chemical Identifiers

CAS 400-98-6
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00007155
InChI Key ATXBGHLILIABGX-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl aniline, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, 4-amino-3-nitrobenzotrifluoride, 4-trifluoromethyl 2-nitroaniline, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, aniline, 2-nitro-4-trifluoromethyl, benzenamine, 2-nitro-4-trifluoromethyl
PubChem CID 67865
IUPAC Name 2-nitro-4-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC104170250
SDS View Documents
Thermo Fisher Scientific
104170250
25g Glass bottle
Each for $69.98
 
Description

Description

Specifications

Chemical Identifiers

400-98-6
206.124
ATXBGHLILIABGX-UHFFFAOYSA-N
67865
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N
C7H5F3N2O2
MFCD00007155
2-nitro-4-trifluoromethyl aniline, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, 4-amino-3-nitrobenzotrifluoride, 4-trifluoromethyl 2-nitroaniline, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, aniline, 2-nitro-4-trifluoromethyl, benzenamine, 2-nitro-4-trifluoromethyl
2-nitro-4-(trifluoromethyl)aniline

Specifications

Authentic
Yellow
25g
97.5
98%
H2NC6H3(NO2)CF3
2-nitro-4-trifluoromethyl aniline, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, 4-amino-3-nitrobenzotrifluoride, 4-trifluoromethyl 2-nitroaniline, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, aniline, 2-nitro-4-trifluoromethyl, benzenamine, 2-nitro-4-trifluoromethyl
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N
206.124
206.12
≥97.5%
Glass bottle
105°C to 109°C
400-98-6
100.0
C7H5F3N2O2
MFCD00007155
ATXBGHLILIABGX-UHFFFAOYSA-N
2-nitro-4-(trifluoromethyl)aniline
67865
Crystals or Crystalline Powder
4-Amino-3-nitrobenzotrifluoride, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-926-3

SDS
Product Certifications