Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-nitrobenzoic acid, 97%

$304.36 - $1,306.36

Chemical Identifiers

Chemical Name or Material 4-Amino-3-nitrobenzoic acid
CAS 1588-83-6
Molecular Formula C7H6N2O4
Formula Weight 182.14
MDL Number MFCD00017009
Beilstein 645206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1924018

  SDS
alfa aesar™
A1924018
50g Each for $304.36

AAA1924030

  SDS
alfa aesar™
A1924030
250g Each for $1,306.36
Description & Specifications

Specifications

Formula Weight 182.14
CAS 1588-83-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 50g
Assay 97%
Beilstein 645206
Molecular Formula C7H6N2O4
Health Hazard 1 Warning
Chemical Name or Material 4-Amino-3-nitrobenzoic acid
TSCA TSCA
Melting Point ≈290°C (dec.)
MDL Number MFCD00017009
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.