missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-methylbenzenesulfonic acid, 98%

$42.72 - $109.59

Chemical Identifiers

CAS 98-33-9
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00025198
InChI Key WQTCZINVPXJNEL-UHFFFAOYSA-N
Synonym 2-amino-5-toluenesulfonic acid, 2-aminotoluene-5-sulfonic acid, 3-methylsulphanilic acid, 4-amino-m-toluenesulfonic acid, 4-amino-m-toluenesulfonic acid so3h=1, 6-aminotoluene-3-sulfonic acid, benzenesulfonic acid, 4-amino-3-methyl, ccris 3412, o-toluidine-m-sulfonic acid, unii-et833cb2ku
PubChem CID 7384
IUPAC Name 4-amino-3-methylbenzenesulfonic acid
SMILES CC1=C(C=CC(=C1)S(=O)(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1987030
View Documents
Alfa Aesar™
A1987030
250g
Each for $42.72
 
AAA198700B
View Documents
Alfa Aesar™
A198700B
1000g
Each for $109.59
 
Description

Description

Specifications

Chemical Identifiers

98-33-9
187.213
WQTCZINVPXJNEL-UHFFFAOYSA-N
7384
CC1=C(C=CC(=C1)S(=O)(=O)O)N
C7H9NO3S
MFCD00025198
2-amino-5-toluenesulfonic acid, 2-aminotoluene-5-sulfonic acid, 3-methylsulphanilic acid, 4-amino-m-toluenesulfonic acid, 4-amino-m-toluenesulfonic acid so3h=1, 6-aminotoluene-3-sulfonic acid, benzenesulfonic acid, 4-amino-3-methyl, ccris 3412, o-toluidine-m-sulfonic acid, unii-et833cb2ku
4-amino-3-methylbenzenesulfonic acid

Specifications

Odorless
250g
C7H9NO3S
2-amino-5-toluenesulfonic acid, 2-aminotoluene-5-sulfonic acid, 3-methylsulphanilic acid, 4-amino-m-toluenesulfonic acid, 4-amino-m-toluenesulfonic acid so3h=1, 6-aminotoluene-3-sulfonic acid, benzenesulfonic acid, 4-amino-3-methyl, ccris 3412, o-toluidine-m-sulfonic acid, unii-et833cb2ku
CC1=C(C=CC(=C1)S(=O)(=O)O)N
187.213
187.22
4-Amino-3-methylbenzenesulfonic acid
>300°C
98-33-9
MFCD00025198
WQTCZINVPXJNEL-UHFFFAOYSA-N
4-amino-3-methylbenzenesulfonic acid
7384
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c

H317-H319

EINECSNumber : 202-658-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS