missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-3-methoxybenzoic acid, 95%, Thermo Scientific Chemicals
$132.67 - $419.19
Chemical Identifiers
CAS | 2486-69-3 |
---|---|
Molecular Formula | C8H9NO3 |
Molecular Weight (g/mol) | 167.164 |
MDL Number | MFCD00016539 |
InChI Key | JNFGLYJROFAOQP-UHFFFAOYSA-N |
Synonym | 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid |
PubChem CID | 288057 |
IUPAC Name | 4-amino-3-methoxybenzoic acid |
SMILES | COC1=C(C=CC(=C1)C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2809803
|
Thermo Scientific Chemicals
H2809803 |
1 g |
Each for $132.67
|
|
|||||
AAH2809806
|
Thermo Scientific Chemicals
H2809806 |
5 g |
Each for $419.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2486-69-3 | |
167.164 | |
JNFGLYJROFAOQP-UHFFFAOYSA-N | |
288057 | |
COC1=C(C=CC(=C1)C(=O)O)N |
C8H9NO3 | |
MFCD00016539 | |
4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid | |
4-amino-3-methoxybenzoic acid |
Specifications
2486-69-3 | |
C8H9NO3 | |
1 g | |
JNFGLYJROFAOQP-UHFFFAOYSA-N | |
4-amino-3-methoxybenzoic acid | |
288057 | |
95% |
186°C to 188°C | |
MFCD00016539 | |
4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid | |
COC1=C(C=CC(=C1)C(=O)O)N | |
167.164 | |
167.16 | |
4-Amino-3-methoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only