missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-hydroxybenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1020722

 View more versions of this product

Catalog No. AAA1020722



Description

Description

Applications
4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

Solubility
Soluble in chloroform, and aqeous ethanol, water (Partly miscible)

Notes
Store at 4°C. Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications

Specifications

4-Amino-3-hydroxybenzoic acid
100g
C7H7NO3
509859
NFPYJDZQOKCYIE-UHFFFAOYSA-N
4-amino-3-hydroxybenzoic acid
137566
98%
~215°C (decomposition)
2374-03-0
MFCD00017094
3-hydroxy-4-aminobenzoic acid, 4-amino-3-hydroxy benzoic acid, benzoic acid, 4-amino-3-hydroxy, chembl1462, 2hdr, pubchem14418, 2-amino-5-carboxyphenol, intermediates-zcf02635, acmc-209g5k, 4-carboxy-2-hydroxyaniline
C1=CC(=C(C=C1C(=O)O)O)N
153.137
153.14

Chemical Identifiers

2374-03-0
153.137
NFPYJDZQOKCYIE-UHFFFAOYSA-N
137566
C1=CC(=C(C=C1C(=O)O)O)N
C7H7NO3
MFCD00017094
3-hydroxy-4-aminobenzoic acid, 4-amino-3-hydroxy benzoic acid, benzoic acid, 4-amino-3-hydroxy, chembl1462, 2hdr, pubchem14418, 2-amino-5-carboxyphenol, intermediates-zcf02635, acmc-209g5k, 4-carboxy-2-hydroxyaniline
4-amino-3-hydroxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents