Please login to your online account to display your discounted pricing

4-Amino-3-hydroxy-1-naphthalenesulfonic Acid, Reagent ACS 90+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  400851000

 View more versions of this product

Catalog No. AC400851000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Powder
Formula Weight 239.25
Quantity 100g
Color Light purple
Assay ≥90.0%
EINECS Number 204-147-3
TSCA TSCA
Solubility Solubility in water: insoluble. Other solubilities: Soluble in hot sodium bisulfite sol., Soluble in alkaline solutions, insoluble in alcohol, ether and benzene
RTECS Number QK1292000
Beilstein 14, 846
Merck Index 15, 450
Synonym 1-Amino-2-naphthol-4-sulfonic acid
Packaging Glass bottle
Sulfate ≤0.2%
Assay Percent Range 90+%
Linear Formula H2NC10H5(OH)SO3H
Health Hazard 1 LIGHT SENSITIVE
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Molecular Formula C10H9NO4S
Residue after Ignition ≤0.1%
Sensitivity Pass Test (To Phosphates)
Name Note 98+%
MDL Number MFCD00004019
CAS 116-63-2
Infrared Spectrum Conforms to Ref Report
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Name Note 98+%
CAS 116-63-2
Purity Grade ACS Reagent Grade
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C10H9NO4S
Linear Formula H2NC10H5(OH)SO3H
Formula Weight 239.25
MDL Number MFCD00004019
Synonym 1-Amino-2-naphthol-4-sulfonic acid
Accessories

LIGHT SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents