missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 400850250

 View more versions of this product

Catalog No. AC400850250



Description

Description

Specifications

Specifications

4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Conforms to Ref Report
0.1% max
0.2% max
295°C
98+%
90+%
H2NC10H5(OH)SO3H
14, 846
1,2,4-acid, 1,2,4-aminonaphtholsulfonic acid, 1-amino-2-naphthol-4-sulfonic acid, 1-amino-4-sulfo-2-naphthol, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 2-hydroxy-4-sulfo-1-naphthylamine, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, unii-w46sk3p33g
RXCMFQDTWCCLBL-UHFFFAOYSA-N
4-amino-3-hydroxynaphthalene-1-sulfonic acid
8316
239.25
90+%
≤0.1%
Glass bottle
Pass Test (To Phosphates)
Purple
25g
116-63-2
C10H9NO4S
MFCD00004019
15, 450
(in sodium carbonate) Passes test
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
239.25
CHEBI:19024
Powder
ACS Reagent

Chemical Identifiers

116-63-2
239.25
RXCMFQDTWCCLBL-UHFFFAOYSA-N
8316
4-amino-3-hydroxynaphthalene-1-sulfonic acid
C10H9NO4S
MFCD00004019
1,2,4-acid, 1,2,4-aminonaphtholsulfonic acid, 1-amino-2-naphthol-4-sulfonic acid, 1-amino-4-sulfo-2-naphthol, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 2-hydroxy-4-sulfo-1-naphthylamine, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, unii-w46sk3p33g
CHEBI:19024
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-147-3

RTECSNumber : QK1292000

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only