missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-3-fluorophenylboronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444500050
Specifications
4-Amino-3-fluorophenylboronic acid pinacol ester | |
5g | |
96% min. (GC) | |
2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorophenylboronicacidpinacolester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
237.08 | |
237.08 |
101°C to 102°C | |
819058-34-9 | |
C12H17BFNO2 | |
AIXGNRNTXUKZLC-UHFFFAOYSA-N | |
17906172 | |
97% |
Chemical Identifiers
819058-34-9 | |
237.08 | |
2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorophenylboronicacidpinacolester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
C12H17BFNO2 | |
AIXGNRNTXUKZLC-UHFFFAOYSA-N | |
17906172 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning