missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-3-fluorophenylboronic acid hydrochloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445280010
Specifications
4-Amino-3-fluorophenylboronic acid hydrochloride | |
Glass Bottle | |
1256355-32-4 | |
C6H7BFNO2·ClH | |
NXCNVCKJELIVDM-UHFFFAOYSA-N | |
46738020 | |
97% |
18 to 19% | |
1g | |
96% min. (HPLC) | |
4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, 4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid,hydrochloride, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1 | |
191.4 | |
191.4 |
Chemical Identifiers
1256355-32-4 | |
191.4 | |
4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, 4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid,hydrochloride, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1 |
C6H7BFNO2·ClH | |
NXCNVCKJELIVDM-UHFFFAOYSA-N | |
46738020 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning