missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-fluorophenylboronic acid hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 1256355-32-4
Molecular Formula C6H8BClFNO2
Molecular Weight (g/mol) 191.39
InChI Key NXCNVCKJELIVDM-UHFFFAOYSA-N
Synonym 4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid,hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
PubChem CID 46738020
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445280010 SDS View Documents Acros Organics
AC445280010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445280050 SDS View Documents Acros Organics
AC445280050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1256355-32-4
191.39
4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid,hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
C6H8BClFNO2
NXCNVCKJELIVDM-UHFFFAOYSA-N
46738020

Specifications

1g
97%
4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid,hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
191.39
191.4
1256355-32-4
C6H8BClFNO2
NXCNVCKJELIVDM-UHFFFAOYSA-N
46738020
4-Amino-3-fluorophenylboronic acid hydrochloride
Safety and Handling

Safety and Handling

Warning