Learn More
4-Amino-3-ethylbenzonitrile, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1073206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Amino-3-ethylbenzonitrile | |
60°C to 66°C | |
MFCD00041431 | |
UN3439 | |
4-amino-3-ethyl-benzonitrile, 4-cyano-2-ethylaniline, 4-amino-3-ethyl benzonitrile, buttpark 13\03-08, benzonitrile, 4-amino-3-ethyl-9ci, 4-amino-3-ethylbenzenecarbonitrile, pubchem16098, pubchem21039, acmc-209xcd, intermediates-zcf02269 | |
CCC1=C(C=CC(=C1)C#N)N | |
146.193 | |
146.19 |
170230-87-2 | |
C9H10N2 | |
5 g | |
7526452 | |
YOHLABDNVQLZIA-UHFFFAOYSA-N | |
4-amino-3-ethylbenzonitrile | |
2735330 | |
96% |
Chemical Identifiers
170230-87-2 | |
146.193 | |
YOHLABDNVQLZIA-UHFFFAOYSA-N | |
2735330 | |
CCC1=C(C=CC(=C1)C#N)N |
C9H10N2 | |
MFCD00041431 | |
4-amino-3-ethyl-benzonitrile, 4-cyano-2-ethylaniline, 4-amino-3-ethyl benzonitrile, buttpark 13\03-08, benzonitrile, 4-amino-3-ethyl-9ci, 4-amino-3-ethylbenzenecarbonitrile, pubchem16098, pubchem21039, acmc-209xcd, intermediates-zcf02269 | |
4-amino-3-ethylbenzonitrile |
Safety and Handling
GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
H302+H332-H311
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only