missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-chloropyridine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3109303

 View more versions of this product

Catalog No. AAH3109303



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Amino-3-chloropyridine
49°C to 53°C
C5H5ClN2
1 g
KJBKPRMEMJKXDV-UHFFFAOYSA-N
3-chloropyridin-4-amine
581853
97%
19798-77-7
Unpleasant
MFCD03984473
4-amino-3-chloropyridine, 3-chloro-pyridin-4-ylamine, 3-chloro-4-pyridinamine, 3-chloro-4-aminopyridine, 3-chloro-4-pyridinylamine, 4-pyridinamine, 3-chloro, 3-chloro-4-pyridinamin, 3-chloro-4-pyridylamine, 4-amino-3-chloro-pyridine, pubchem6653
C1=CN=CC(=C1N)Cl
128.559
128.56

Chemical Identifiers

19798-77-7
128.559
KJBKPRMEMJKXDV-UHFFFAOYSA-N
581853
C1=CN=CC(=C1N)Cl
C5H5ClN2
MFCD03984473
4-amino-3-chloropyridine, 3-chloro-pyridin-4-ylamine, 3-chloro-4-pyridinamine, 3-chloro-4-aminopyridine, 3-chloro-4-pyridinylamine, 4-pyridinamine, 3-chloro, 3-chloro-4-pyridinamin, 3-chloro-4-pyridylamine, 4-amino-3-chloro-pyridine, pubchem6653
3-chloropyridin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only