Please login to your online account to display your discounted pricing

4-Amino-3-chlorophenol hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Amino-3-chlorophenol hydrochloride
Assay Percent Range 98%
Molecular Formula C6H6ClNO·HCl
Linear Formula ClC6H3(OH)NH2·HCl
Formula Weight 180.03
MDL Number MFCD00143110
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399310050

  SDS
thermo fisher scientific
AC399310050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399310250

  SDS
thermo fisher scientific
AC399310250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.03
Quantity 5g
Chemical Name or Material 4-Amino-3-chlorophenol hydrochloride
Molecular Formula C6H6ClNO·HCl
Linear Formula ClC6H3(OH)NH2·HCl
MDL Number MFCD00143110
CAS 52671-64-4
Assay Percent Range 98%