Please login to your online account to display your discounted pricing

4-Amino-3-chlorophenol hydrochloride, 98%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC399310250

 View more versions of this product

Catalog No. AC399310250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.03
Quantity 25g
Assay Percent Range 98%
Linear Formula ClC6H3(OH)NH2·HCl
Chemical Name or Material 4-Amino-3-chlorophenol hydrochloride
Molecular Formula C6H6ClNO·HCl
CAS 52671-64-4
MDL Number MFCD00143110

Specifications

Chemical Name or Material 4-Amino-3-chlorophenol hydrochloride
Assay Percent Range 98%
Molecular Formula C6H6ClNO·HCl
Linear Formula ClC6H3(OH)NH2·HCl
Formula Weight 180.03
MDL Number MFCD00143110