missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-chlorophenol, 95%

$145.95 - $436.68

Chemical Identifiers

CAS 17609-80-2
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00801116
InChI Key PNLPXABQLXSICH-UHFFFAOYSA-N
Synonym 2-chloro-4-hydroxyaniline, 3-13-00-01184 beilstein handbook reference, 3-chloro-4-aminophenol, 3-chloro-4-aminophenol;4-amino-3-chloro-pheno, 4-amino-3-chloro-pheno, 4-amino-3-chloro-phenol, amino 4--3-chlorophenol, l-valine ethyl ester hydrochloride; ethyl 2-amino-3-methylbutanoate hydrochloride, phenol, 4-amino-3-chloro, pubchem4513
PubChem CID 87184
IUPAC Name 4-amino-3-chlorophenol
SMILES C1=CC(=C(C=C1O)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66310MD
View Documents
Alfa Aesar™
H66310MD
250mg Each for $145.95
 
AAH6631003
View Documents
Alfa Aesar™
H6631003
1g Each for $436.68
 
Specifications

Chemical Identifiers

17609-80-2
143.57
PNLPXABQLXSICH-UHFFFAOYSA-N
87184
C1=CC(=C(C=C1O)Cl)N
C6H6ClNO
MFCD00801116
2-chloro-4-hydroxyaniline, 3-13-00-01184 beilstein handbook reference, 3-chloro-4-aminophenol, 3-chloro-4-aminophenol;4-amino-3-chloro-pheno, 4-amino-3-chloro-pheno, 4-amino-3-chloro-phenol, amino 4--3-chlorophenol, l-valine ethyl ester hydrochloride; ethyl 2-amino-3-methylbutanoate hydrochloride, phenol, 4-amino-3-chloro, pubchem4513
4-amino-3-chlorophenol

Specifications

White
250mg
C6H6ClNO
UN2811
2-chloro-4-hydroxyaniline, 3-13-00-01184 beilstein handbook reference, 3-chloro-4-aminophenol, 3-chloro-4-aminophenol;4-amino-3-chloro-pheno, 4-amino-3-chloro-pheno, 4-amino-3-chloro-phenol, amino 4--3-chlorophenol, l-valine ethyl ester hydrochloride; ethyl 2-amino-3-methylbutanoate hydrochloride, phenol, 4-amino-3-chloro, pubchem4513
C1=CC(=C(C=C1O)Cl)N
143.57
143.57
95%
159°C to 160°C
17609-80-2
MFCD00801116
2802558
PNLPXABQLXSICH-UHFFFAOYSA-N
4-amino-3-chlorophenol
87184
Crystalline Powder
4-Amino-3-chlorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 241-583-3

RTECSNumber : SJ5710000

TSCA : No

Recommended Storage : Ambient temperatures