missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-chlorobenzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1215414

 View more versions of this product

Catalog No. AAA1215414



Specifications

Specifications

4-Amino-3-chlorobenzonitrile
25g
C7H5ClN2
UN3439
3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04
C1=CC(=C(C=C1C#N)Cl)N
152.581
152.58
98°C to 104°C
21803-75-8
MFCD00052917
1817655
OREVCMGFYSUYPX-UHFFFAOYSA-N
4-amino-3-chlorobenzonitrile
519896
98%

Chemical Identifiers

21803-75-8
152.581
OREVCMGFYSUYPX-UHFFFAOYSA-N
519896
C1=CC(=C(C=C1C#N)Cl)N
C7H5ClN2
MFCD00052917
3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04
4-amino-3-chlorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents