Please login to your online account to display your discounted pricing

4-Amino-3-chloro-5-nitrobenzotrifluoride, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350070050

 View more versions of this product

Catalog No. AC350070050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 240.57
Quantity 5g
Color Yellow to orange-brown
Melting Point 63° to 66°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00042153
Molecular Formula C7 H4 Cl F3 N2 O2
CAS 57729-79-0
CAS Min % 98.5
Chemical Name or Material 4-Amino-3-chloro-5-nitrobenzotrifluoride
Assay Percent Range 99%
Name Note 99%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Amino-3-chloro-5-nitrobenzotrifluoride
Name Note 99%
CAS 57729-79-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7 H4 Cl F3 N2 O2
Formula Weight 240.57
MDL Number MFCD00042153
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.

SDS & Safety Documents