missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-chloro-5-nitrobenzotrifluoride, 99%, ACROS Organics™

Chemical Identifiers

CAS 57729-79-0
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7 H4 Cl F3 N2 O2
Formula Weight 240.57
MDL Number MFCD00042153
Chemical Name or Material 4-Amino-3-chloro-5-nitrobenzotrifluoride, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC350070050 SDS Thermo Fisher Scientific
350070050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57729-79-0
100.0
C7 H4 Cl F3 N2 O2
MFCD00042153
≥98.5%
98.5
99%
240.57
4-Amino-3-chloro-5-nitrobenzotrifluoride, 99%

Specifications

Crystalline Powder
5g
63° to 66°C
Authentic
C7 H4 Cl F3 N2 O2
98.5
Glass bottle
≥98.5%
240.57
Yellow to orange-brown
100.0
MFCD00042153
57729-79-0
99%
4-Amino-3-chloro-5-nitrobenzotrifluoride, 99%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning