Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-chloro-5-methylbenzonitrile, tech. 90%

Chemical Identifiers

Chemical Name or Material 4-Amino-3-chloro-5-methylbenzonitrile
CAS 158296-69-6
Purity Grade Technical Grade
Molecular Formula C8H7ClN2
Formula Weight 166.61
MDL Number MFCD00041476
UN Number UN3439
Beilstein 7527075
Synonym 2-Chloro-4-cyano-6-methylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1254103

  SDS
alfa aesar™
L1254103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1254106

  SDS
alfa aesar™
L1254106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.61
Chemical Name or Material 4-Amino-3-chloro-5-methylbenzonitrile
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Assay 90%
Molecular Formula C8H7ClN2
MDL Number MFCD00041476
CAS 158296-69-6
UN Number UN3439
Synonym 2-Chloro-4-cyano-6-methylaniline
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 128° to 130°C
Purity Grade Technical Grade
Beilstein 7527075
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.