missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-chloro-5-methylbenzonitrile, tech. 90%

Manufacturer:  Alfa Aesar™ L1254103

 View more versions of this product

Catalog No. AAL1254103



Specifications

Specifications

4-Amino-3-chloro-5-methylbenzonitrile
1g
C8H7ClN2
UN3439
4-amino-3-chloro-5-methyl-benzonitrile, benzonitrile, 4-amino-3-chloro-5-methyl, 4-amino-3-chloro-5-methylbenzenecarbonitrile, 2-chloro-4-cyano-6-methylaniline, acmc-1cuhq, ksc498c2h, 2-chloro-4-cyano-6-methylaniline, 4-amino-3-chloro-5-methyl benzonitrile, 4-azanyl-3-chloranyl-5-methyl-benzenecarbonitrile
CC1=C(C(=CC(=C1)C#N)Cl)N
166.608
166.61
Technical
128°C to 130°C
158296-69-6
MFCD00041476
7527075
NDTNVCCDQAOBSZ-UHFFFAOYSA-N
4-amino-3-chloro-5-methylbenzonitrile
2735301
90%

Chemical Identifiers

158296-69-6
166.608
NDTNVCCDQAOBSZ-UHFFFAOYSA-N
2735301
CC1=C(C(=CC(=C1)C#N)Cl)N
C8H7ClN2
MFCD00041476
4-amino-3-chloro-5-methyl-benzonitrile, benzonitrile, 4-amino-3-chloro-5-methyl, 4-amino-3-chloro-5-methylbenzenecarbonitrile, 2-chloro-4-cyano-6-methylaniline, acmc-1cuhq, ksc498c2h, 2-chloro-4-cyano-6-methylaniline, 4-amino-3-chloro-5-methyl benzonitrile, 4-azanyl-3-chloranyl-5-methyl-benzenecarbonitrile
4-amino-3-chloro-5-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents