missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-bromopyridine, 98%

$78.91 - $78.91

Chemical Identifiers

CAS 13534-98-0
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD02068297
InChI Key DDQYSZWFFXOXER-UHFFFAOYSA-N
Synonym 3-bromo-4-aminopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-pyridineamine, 3-bromo-4-pyridinylamine, 3-bromo-4-pyridylamine, 3-bromo-pyridin-4-ylamine, 4-amino-3-bromopyridine, 4-pyridinamine, 3-bromo, acmc-1bnz6, pubchem1242
PubChem CID 26095
IUPAC Name 3-bromopyridin-4-amine
SMILES C1=CN=CC(=C1N)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1952103
View Documents
Alfa Aesar™
L1952103
1g Each for $78.91
 
Specifications

Chemical Identifiers

13534-98-0
173.013
DDQYSZWFFXOXER-UHFFFAOYSA-N
26095
C1=CN=CC(=C1N)Br
C5H5BrN2
MFCD02068297
3-bromo-4-aminopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-pyridineamine, 3-bromo-4-pyridinylamine, 3-bromo-4-pyridylamine, 3-bromo-pyridin-4-ylamine, 4-amino-3-bromopyridine, 4-pyridinamine, 3-bromo, acmc-1bnz6, pubchem1242
3-bromopyridin-4-amine

Specifications

65°C to 69°C
13534-98-0
MFCD02068297
3-bromo-4-aminopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-pyridineamine, 3-bromo-4-pyridinylamine, 3-bromo-4-pyridylamine, 3-bromo-pyridin-4-ylamine, 4-amino-3-bromopyridine, 4-pyridinamine, 3-bromo, acmc-1bnz6, pubchem1242
C1=CN=CC(=C1N)Br
173.013
173.01
4-Amino-3-bromopyridine
1g
C5H5BrN2
110183
DDQYSZWFFXOXER-UHFFFAOYSA-N
3-bromopyridin-4-amine
26095
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures