Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-bromopyridine, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1952106

 View more versions of this product

Catalog No. AAL1952106

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 173.01
Chemical Name or Material 4-Amino-3-bromopyridine
CAS 13534-98-0
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Melting Point 65° to 69°C
Molecular Formula C5H5BrN2
Quantity 5g
MDL Number MFCD02068297
Synonym 3-Bromo-4-pyridinamine
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 110183

Specifications

Chemical Name or Material 4-Amino-3-bromopyridine
CAS 13534-98-0
Molecular Formula C5H5BrN2
Formula Weight 173.01
MDL Number MFCD02068297
Beilstein 110183
Synonym 3-Bromo-4-pyridinamine
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.