missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-bromobenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H55483MD

 View more versions of this product

Catalog No. AAH55483MD



Specifications

Specifications

4-Amino-3-bromobenzonitrile
250mg
C7H5BrN2
UN3439
POESQIHWIIWNJL-UHFFFAOYSA-N
4-amino-3-bromobenzonitrile
1515279
97%
106°C to 110°C
50397-74-5
MFCD01861386
2-bromo-4-cyanoaniline
C1=CC(=C(C=C1C#N)Br)N
197.035
197.03

Chemical Identifiers

50397-74-5
197.035
POESQIHWIIWNJL-UHFFFAOYSA-N
1515279
C1=CC(=C(C=C1C#N)Br)N
C7H5BrN2
MFCD01861386
2-bromo-4-cyanoaniline
4-amino-3-bromobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302+H312+H332-H315-H317-H319-H335-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents