missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-bromobenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5548306

 View more versions of this product

Catalog No. AAH5548306



Specifications

Specifications

4-Amino-3-bromobenzonitrile
5g
C7H5BrN2
UN3439
POESQIHWIIWNJL-UHFFFAOYSA-N
4-amino-3-bromobenzonitrile
1515279
97%
106°C to 110°C
50397-74-5
MFCD01861386
2-bromo-4-cyanoaniline
C1=CC(=C(C=C1C#N)Br)N
197.035
197.03

Chemical Identifiers

50397-74-5
197.035
POESQIHWIIWNJL-UHFFFAOYSA-N
1515279
C1=CC(=C(C=C1C#N)Br)N
C7H5BrN2
MFCD01861386
2-bromo-4-cyanoaniline
4-amino-3-bromobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents