Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-bromo-5-iodopyridine, 99%

Chemical Identifiers

Chemical Name or Material 4-Amino-3-bromo-5-iodopyridine
CAS 902837-39-2
Molecular Formula C5H4BrIN2
Formula Weight 298.91g/mol
MDL Number MFCD07781147
Synonym 3-Bromo-5-iodo-4-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50030MD

 
alfa aesar™
H50030-MD
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 250mg
Chemical Name or Material 4-Amino-3-bromo-5-iodopyridine
CAS 902837-39-2
Assay 99%
Molecular Formula C5H4BrIN2
Formula Weight 298.91g/mol
MDL Number MFCD07781147
Synonym 3-Bromo-5-iodo-4-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.