Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3,5-dimethyl-1,2,4-triazole, 98%

$49.11 - $186.84

Chemical Identifiers

Chemical Name or Material 4-Amino-3,5-dimethyl-1,2,4-triazole
CAS 3530-15-2
Molecular Formula C4H8N4
Formula Weight 112.14
MDL Number MFCD00051647
Beilstein 115482
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2041703

  SDS
alfa aesar™
B2041703
1g Each for $49.11

AAB2041706

  SDS
alfa aesar™
B2041706
5g Each for $186.84
Description & Specifications

Specifications

Formula Weight 112.14
Chemical Name or Material 4-Amino-3,5-dimethyl-1,2,4-triazole
Melting Point 196° to 199°C
Assay 98%
Quantity 1g
Molecular Formula C4H8N4
MDL Number MFCD00051647
CAS 3530-15-2
Beilstein 115482
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.