missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3,5-diiodobenzoic acid, tech. 90%

$50.12 - $181.97

Chemical Identifiers

CAS 2122-61-4
Molecular Formula C7H5I2NO2
Molecular Weight (g/mol) 388.931
MDL Number MFCD00007684
InChI Key WXTVPMWCUMEVSZ-UHFFFAOYSA-N
Synonym benzoic acid, 4-amino-3,5-diiodo, 3,5-dijod-4-aminohippursaeure, 3,5-diiodo-4-aminobenzoic acid, 3,5-dijod-4-aminohippursaeure german, 4-amino-3,5-diiodo-benzoicacid, pubchem4681, acmc-1cq3z, wln: zr bi fi dvq, labotest-bb lt00080707, 4-amino-3,5-diiodo-benzoicaci
PubChem CID 16460
IUPAC Name 4-amino-3,5-diiodobenzoic acid
SMILES C1=C(C=C(C(=C1I)N)I)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1061706 View Documents Alfa Aesar™
A1061706
5g Each for $50.12
 
AAA1061714 View Documents Alfa Aesar™
A1061714
25g Each for $181.97
 
Description

Description

Applications
4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.

Solubility
Soluble in water (partly miscible).

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications

Chemical Identifiers

2122-61-4
388.931
WXTVPMWCUMEVSZ-UHFFFAOYSA-N
16460
C1=C(C=C(C(=C1I)N)I)C(=O)O
C7H5I2NO2
MFCD00007684
benzoic acid, 4-amino-3,5-diiodo, 3,5-dijod-4-aminohippursaeure, 3,5-diiodo-4-aminobenzoic acid, 3,5-dijod-4-aminohippursaeure german, 4-amino-3,5-diiodo-benzoicacid, pubchem4681, acmc-1cq3z, wln: zr bi fi dvq, labotest-bb lt00080707, 4-amino-3,5-diiodo-benzoicaci
4-amino-3,5-diiodobenzoic acid

Specifications

>300°C
2122-61-4
MFCD00007684
benzoic acid, 4-amino-3,5-diiodo, 3,5-dijod-4-aminohippursaeure, 3,5-diiodo-4-aminobenzoic acid, 3,5-dijod-4-aminohippursaeure german, 4-amino-3,5-diiodo-benzoicacid, pubchem4681, acmc-1cq3z, wln: zr bi fi dvq, labotest-bb lt00080707, 4-amino-3,5-diiodo-benzoicaci
C1=C(C=C(C(=C1I)N)I)C(=O)O
388.931
388.93
Technical
5g
C7H5I2NO2
2103037
WXTVPMWCUMEVSZ-UHFFFAOYSA-N
4-amino-3,5-diiodobenzoic acid
16460
90%
4-Amino-3,5-diiodobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG2310000