Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3,5-dichlorobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Amino-3,5-dichlorobenzonitrile
CAS 78473-00-4
Molecular Formula C7H4Cl2N2
Formula Weight 187.03
MDL Number MFCD00017341
UN Number UN3439
Beilstein 2719605
Synonym 4-Cyano-2,6-dichloroaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1126906

  SDS
alfa aesar™
A1126906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1126903

  SDS
alfa aesar™
A1126903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1126914

  SDS
alfa aesar™
A1126914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.03
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 78473-00-4
Synonym 4-Cyano-2,6-dichloroaniline
UN Number UN3439
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H4Cl2N2
Beilstein 2719605
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 117° to 119°C
Chemical Name or Material 4-Amino-3,5-dichlorobenzonitrile
Assay 97%
Health Hazard 1 Danger
MDL Number MFCD00017341
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.