missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3,5-dichlorobenzoic acid +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC394070010

 View more versions of this product

Catalog No. AC394070010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.0%
206.03
Cream
98.0
NH2C6H2(Cl)2CO2H
C7H5Cl2NO2
Authentic
98+%
Powder
1g
100.0
56961-25-2
Glass bottle
MFCD00017091
4-Amino-3, 5-dichlorobenzoic acid

Chemical Identifiers

56961-25-2
100.0
C7H5Cl2NO2
206.03
4-Amino-3, 5-dichlorobenzoic acid
98.0
98+%
NH2C6H2(Cl)2CO2H
MFCD00017091
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.