Please login to your online account to display your discounted pricing

4-Amino-3,5-dichlorobenzoic acid +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Amino-3, 5-dichlorobenzoic acid
CAS 56961-25-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H5Cl2NO2
Linear Formula NH2C6H2(Cl)2CO2H
Formula Weight 206.03
MDL Number MFCD00017091
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC394070010

  SDS
thermo fisher scientific
AC394070010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Powder
Formula Weight 206.03
Quantity 1g
Color Cream
CAS Max % 100.0
CAS Min % 98.0
CAS 56961-25-2
Linear Formula NH2C6H2(Cl)2CO2H
Packaging Glass bottle
Molecular Formula C7H5Cl2NO2
MDL Number MFCD00017091
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Chemical Name or Material 4-Amino-3, 5-dichlorobenzoic acid
Assay Percent Range 98+%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.