missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2-(trifluoromethyl)benzonitrile, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442470010

 View more versions of this product

Catalog No. AC442470010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Amino-2-(trifluoromethyl)benzonitrile
Glass bottle
144.0°C to 146.0°C
654-70-6
100.0
C8H5F3N2
PMDYLCUKSLBUHO-UHFFFAOYSA-N
4-amino-2-(trifluoromethyl)benzonitrile
522170
Powder
Authentic
White
1g
96.0
96% min. (HPLC)
2-cyano-5-aminobenzotrifluoride, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino 2-trifluoromethyl benzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile, 4-amino-2-trifluoromethyl benzonitrile, 4-cyano-3-trifluoromethyl aniline, 4-cyano-3-trifluoromethylaniline, 5-amino-2-cyanobenzotrifluoride, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08
C1=CC(=C(C=C1N)C(F)(F)F)C#N
186.14
186.14
97%

Chemical Identifiers

654-70-6
186.14
2-cyano-5-aminobenzotrifluoride, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino 2-trifluoromethyl benzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile, 4-amino-2-trifluoromethyl benzonitrile, 4-cyano-3-trifluoromethyl aniline, 4-cyano-3-trifluoromethylaniline, 5-amino-2-cyanobenzotrifluoride, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08
4-amino-2-(trifluoromethyl)benzonitrile
C8H5F3N2
PMDYLCUKSLBUHO-UHFFFAOYSA-N
522170
C1=CC(=C(C=C1N)C(F)(F)F)C#N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS Documents