missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 654-70-6 |
---|---|
Molecular Formula | C8H5F3N2 |
Molecular Weight (g/mol) | 186.14 |
InChI Key | PMDYLCUKSLBUHO-UHFFFAOYSA-N |
Synonym | 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile |
PubChem CID | 522170 |
IUPAC Name | 4-amino-2-(trifluoromethyl)benzonitrile |
SMILES | C1=CC(=C(C=C1N)C(F)(F)F)C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC442470010
|
Thermo Scientific Chemicals
442470010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC442470050
|
Thermo Scientific Chemicals
442470050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
654-70-6 | |
186.14 | |
4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile | |
4-amino-2-(trifluoromethyl)benzonitrile |
C8H5F3N2 | |
PMDYLCUKSLBUHO-UHFFFAOYSA-N | |
522170 | |
C1=CC(=C(C=C1N)C(F)(F)F)C#N |
Specifications
654-70-6 | |
100.0 | |
96% min. (HPLC) | |
C8H5F3N2 | |
4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile | |
C1=CC(=C(C=C1N)C(F)(F)F)C#N | |
186.14 | |
186.14 | |
4-Amino-2-(trifluoromethyl)benzonitrile |
96.0 | |
Authentic | |
Glass bottle | |
1g | |
PMDYLCUKSLBUHO-UHFFFAOYSA-N | |
4-amino-2-(trifluoromethyl)benzonitrile | |
522170 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
RUO â Research Use Only