missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-(trifluoromethyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ L12346MD

 View more versions of this product

Catalog No. AAL12346MD



Specifications

Specifications

4-Amino-2-(trifluoromethyl)benzonitrile
250mg
C8H5F3N2
UN3439
4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile
C1=CC(=C(C=C1N)C(F)(F)F)C#N
186.137
186.14
136°C to 139°C
654-70-6
MFCD00042155
3278074
PMDYLCUKSLBUHO-UHFFFAOYSA-N
4-amino-2-(trifluoromethyl)benzonitrile
522170
97%

Chemical Identifiers

654-70-6
186.137
PMDYLCUKSLBUHO-UHFFFAOYSA-N
522170
C1=CC(=C(C=C1N)C(F)(F)F)C#N
C8H5F3N2
MFCD00042155
4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile
4-amino-2-(trifluoromethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents