Learn More
4-Amino-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1234603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Amino-2-(trifluoromethyl)benzonitrile | |
136°C to 139°C | |
MFCD00042155 | |
UN3439 | |
4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile | |
C1=CC(=C(C=C1N)C(F)(F)F)C#N | |
186.137 | |
186.14 |
654-70-6 | |
C8H5F3N2 | |
1 g | |
3278074 | |
PMDYLCUKSLBUHO-UHFFFAOYSA-N | |
4-amino-2-(trifluoromethyl)benzonitrile | |
522170 | |
97% |
Chemical Identifiers
654-70-6 | |
186.137 | |
PMDYLCUKSLBUHO-UHFFFAOYSA-N | |
522170 | |
C1=CC(=C(C=C1N)C(F)(F)F)C#N |
C8H5F3N2 | |
MFCD00042155 | |
4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile | |
4-amino-2-(trifluoromethyl)benzonitrile |
Safety and Handling
GHS H Statement
H301-H311-H331-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only