Please login to your online account to display your discounted pricing

4-Amino-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Amino-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
CAS 142847-17-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16N2O5
Linear Formula (CH3)3COC(O)NHCH(CO2H)CH2C(O)NH2
Formula Weight 232.23
MDL Number MFCD00068248
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00924EA

 
fisher scientific company
SB00924EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.23
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-Amino-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
Assay Percent Range 97%
Linear Formula (CH3)3COC(O)NHCH(CO2H)CH2C(O)NH2
MDL Number MFCD00068248
CAS 142847-17-4
Packaging Amber Glass Bottle
Molecular Formula C9H16N2O5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.