Please login to your online account to display your discounted pricing

4-Amino-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB00924EA

 View more versions of this product

Catalog No. SB00924EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.23
Quantity 10g
Assay 97%
CAS Min % 97.0
Linear Formula (CH3)3COC(O)NHCH(CO2H)CH2C(O)NH2
CAS 142847-17-4
Assay Percent Range 97%
Chemical Name or Material 4-Amino-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
MDL Number MFCD00068248
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Molecular Formula C9H16N2O5

Specifications

Chemical Name or Material 4-Amino-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
CAS 142847-17-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16N2O5
Linear Formula (CH3)3COC(O)NHCH(CO2H)CH2C(O)NH2
Formula Weight 232.23
MDL Number MFCD00068248
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents