missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-methylpyridine, 98%

Manufacturer:  Alfa Aesar™ H55036MD

 View more versions of this product

Catalog No. AAH55036MD



Description

Description

Applications
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Amino-2-methylpyridine
250mg
C6H8N2
2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-2-methylpyridine, 4-amino-2-picoline, 4-amino-6-picoline, 4-amino-picoline, 4-pyridinamine, 2-methyl, pubchem1118
GNCLPNMQEGMNTG-UHFFFAOYSA-N
2-methylpyridin-4-amine
728670
98%
93°C to 97°C
18437-58-6
MFCD00186509
Soluble in methanol.
CC1=NC=CC(=C1)N
108.144
108.14

Chemical Identifiers

18437-58-6
108.144
GNCLPNMQEGMNTG-UHFFFAOYSA-N
728670
CC1=NC=CC(=C1)N
C6H8N2
MFCD00186509
2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-2-methylpyridine, 4-amino-2-picoline, 4-amino-6-picoline, 4-amino-picoline, 4-pyridinamine, 2-methyl, pubchem1118
2-methylpyridin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents