missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3165703

 View more versions of this product

Catalog No. AAH3165703



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Amino-2-methylbenzoic acid
C8H9NO2
1 g
XRSQZFJLEPBPOZ-UHFFFAOYSA-N
4-amino-2-methylbenzoic acid
241632
98%
2486-75-1
MFCD06656133
4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
CC1=C(C=CC(=C1)N)C(=O)O
151.165
151.16
Solid

Chemical Identifiers

2486-75-1
151.165
XRSQZFJLEPBPOZ-UHFFFAOYSA-N
241632
CC1=C(C=CC(=C1)N)C(=O)O
C8H9NO2
MFCD06656133
4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
4-amino-2-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only