missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-2-mercaptopyrimidine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 214770010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Amino-2-mercaptopyrimidine | |
96.0 | |
Yellow | |
96% min. (HPLC) | |
C4H5N3S | |
2-thiocytosine, 4-amino-2-mercaptopyrimidine, thiocytosine, 4-aminopyrimidine-2-thiol, cytosine, thio, cytosine, 2-thio, 2 1h-pyrimidinethione, 4-amino, 2 1h-pyrimidinone, 4-aminothio, 4-amino-2 1h-pyrimidinethione, 4-amino-2-thiopyrimidine | |
C1=C(NC(=S)N=C1)N | |
127.17 | |
127.17 | |
Powder |
333-49-3 | |
100.0 | |
Authentic | |
Glass bottle | |
1 g | |
DCPSTSVLRXOYGS-UHFFFAOYSA-N | |
6-amino-1H-pyrimidine-2-thione | |
2724245 | |
97% |
Chemical Identifiers
333-49-3 | |
127.17 | |
2-thiocytosine, 4-amino-2-mercaptopyrimidine, thiocytosine, 4-aminopyrimidine-2-thiol, cytosine, thio, cytosine, 2-thio, 2 1h-pyrimidinethione, 4-amino, 2 1h-pyrimidinone, 4-aminothio, 4-amino-2 1h-pyrimidinethione, 4-amino-2-thiopyrimidine | |
6-amino-1H-pyrimidine-2-thione |
C4H5N3S | |
DCPSTSVLRXOYGS-UHFFFAOYSA-N | |
2724245 | |
C1=C(NC(=S)N=C1)N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
EINECSNumber : 206-374-3
RUO – Research Use Only