missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-2-fluorophenylboronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444490050
Specifications
4-Amino-2-fluorophenylboronic acid pinacol ester | |
Brown to White | |
5g | |
97% | |
3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)N)F | |
237.08 | |
237.08 | |
97% |
97 % min. consistent with structure (1H-NMR) | |
163°C to 166°C | |
819057-45-9 | |
C12H17BFNO2 | |
FLMNWVXAEGUVNY-UHFFFAOYSA-N | |
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
44755207 | |
Crystalline Powder |
Chemical Identifiers
819057-45-9 | |
237.08 | |
3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
C12H17BFNO2 | |
FLMNWVXAEGUVNY-UHFFFAOYSA-N | |
44755207 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)N)F |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning