Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-fluorophenol, 97%

$123.55 - $439.97

Chemical Identifiers

Chemical Name or Material 4-Amino-2-fluorophenol
CAS 399-96-2
Molecular Formula C6H6FNO
Formula Weight 127.12
MDL Number MFCD00671760
Synonym 3-Fluoro-4-hydroxyaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2593603

  SDS
alfa aesar™
H2593603
1g Each for $123.55

AAH2593606

  SDS
alfa aesar™
H2593606
5g Each for $439.97
Description & Specifications

Specifications

Formula Weight 127.12
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 399-96-2
Health Hazard 1 Danger
MDL Number MFCD00671760
Melting Point 170° to 172°C
Synonym 3-Fluoro-4-hydroxyaniline
Quantity 1g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Amino-2-fluorophenol
Molecular Formula C6H6FNO
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.