missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 96%

$107.64 - $342.35

Chemical Identifiers

CAS 819057-45-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.081
MDL Number MFCD09951877
InChI Key FLMNWVXAEGUVNY-UHFFFAOYSA-N
Synonym 4-amino-2-fluorophenylboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, 4-amino-2-fluorophenyl boronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline
PubChem CID 44755207
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59916MD View Documents Alfa Aesar™
H59916MD
250mg Each for $107.64
 
AAH5991603 View Documents Alfa Aesar™
H5991603
1g Each for $342.35
 
Specifications

Chemical Identifiers

819057-45-9
237.081
FLMNWVXAEGUVNY-UHFFFAOYSA-N
44755207
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)N)F
C12H17BFNO2
MFCD09951877
4-amino-2-fluorophenylboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, 4-amino-2-fluorophenyl boronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

Specifications

250mg
C12H17BFNO2
4-amino-2-fluorophenylboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, 4-amino-2-fluorophenyl boronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)N)F
237.081
237.08
4-Amino-2-fluorobenzeneboronic acid pinacol ester
819057-45-9
MFCD09951877
FLMNWVXAEGUVNY-UHFFFAOYSA-N
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
44755207
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning