missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
$265.89 - $872.71
Chemical Identifiers
CAS | 1326283-60-6 |
---|---|
Molecular Formula | C13H19BFNO3 |
Molecular Weight (g/mol) | 267.11 |
MDL Number | MFCD22419265 |
InChI Key | JWACNMHKJHEWAQ-UHFFFAOYSA-N |
Synonym | 4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline |
PubChem CID | 67363295 |
IUPAC Name | 5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
SMILES | COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH59944MD
|
Thermo Scientific Chemicals
H59944MD |
250 mg |
Each for $265.89
|
|
|||||
AAH5994403
|
Thermo Scientific Chemicals
H5994403 |
1 g |
Each for $872.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1326283-60-6 | |
267.11 | |
JWACNMHKJHEWAQ-UHFFFAOYSA-N | |
67363295 | |
COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1 |
C13H19BFNO3 | |
MFCD22419265 | |
4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline | |
5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Specifications
1326283-60-6 | |
MFCD22419265 | |
4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline | |
COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1 | |
267.11 | |
267.11 | |
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester |
C13H19BFNO3 | |
250 mg | |
JWACNMHKJHEWAQ-UHFFFAOYSA-N | |
5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
67363295 | |
96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only