missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chloropyrimidine, 98%

Manufacturer:  Alfa Aesar™ H5587503

 View more versions of this product

Catalog No. AAH5587503



Specifications

Specifications

4-Amino-2-chloropyrimidine
1g
C4H4ClN3
2-chloro-4-amino-pyrimidine
C1=CN=C(N=C1N)Cl
129.547
129.55
197°C to 205°C
7461-50-9
MFCD00194051
LPBDZVNGCNTELM-UHFFFAOYSA-N
2-chloropyrimidin-4-amine
345752
98%

Chemical Identifiers

7461-50-9
129.547
LPBDZVNGCNTELM-UHFFFAOYSA-N
345752
C1=CN=C(N=C1N)Cl
C4H4ClN3
MFCD00194051
2-chloro-4-amino-pyrimidine
2-chloropyrimidin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents