Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chloropyrimidine-5-carbonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Amino-2-chloropyrimidine-5-carbonitrile
CAS 94741-69-2
Molecular Formula C5H3ClN4
Formula Weight 154.56
MDL Number MFCD00052343
UN Number UN3439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4395209

  SDS
alfa aesar™
4395209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.56
Physical Form Powder or Flakes
MDL Number MFCD00052343
Chemical Name or Material 4-Amino-2-chloropyrimidine-5-carbonitrile
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 243° to 245°C
Health Hazard 1 Danger
Quantity 10g
UN Number UN3439
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 94741-69-2
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Molecular Formula C5H3ClN4
Assay 97%
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.