missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chloropyrimidine-5-carbonitrile, 97%

Manufacturer:  Alfa Aesar™ 4395209

 View more versions of this product

Catalog No. AA4395209



Specifications

Specifications

4-Amino-2-chloropyrimidine-5-carbonitrile
10g
C5H3ClN4
UN3439
WDHFCSOENXEMRC-UHFFFAOYSA-N
4-amino-2-chloropyrimidine-5-carbonitrile
2801166
Powder or Flakes
243°C to 245°C
94741-69-2
MFCD00052343
4-amino-2-chloro-5-pyrimidinecarbonitrile, 5-pyrimidinecarbonitrile, 4-amino-2-chloro, 4-amino-2-chloro-pyrimidine-5-carbonitrile, pubchem6934, kinome_3477, acmc-209rte, buttpark 6040-60, aminochloropyrimidinecarbonitrile, 2-chloro-4-amino-5-cyanopyrimidine, 4-amino-2-chloro-5-cyanopyrimidine
C1=C(C(=NC(=N1)Cl)N)C#N
154.557
154.56
97%

Chemical Identifiers

94741-69-2
154.557
WDHFCSOENXEMRC-UHFFFAOYSA-N
2801166
C1=C(C(=NC(=N1)Cl)N)C#N
C5H3ClN4
MFCD00052343
4-amino-2-chloro-5-pyrimidinecarbonitrile, 5-pyrimidinecarbonitrile, 4-amino-2-chloro, 4-amino-2-chloro-pyrimidine-5-carbonitrile, pubchem6934, kinome_3477, acmc-209rte, buttpark 6040-60, aminochloropyrimidinecarbonitrile, 2-chloro-4-amino-5-cyanopyrimidine, 4-amino-2-chloro-5-cyanopyrimidine
4-amino-2-chloropyrimidine-5-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents